Assay Details
Gene Symbol: Cenpn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008366.1 | NP_001008367.1 | 3 - 4 | 388 | 98 | |
XM_006255686.2 | XP_006255748.1 | 3 - 4 | 502 | 98 | ||
XM_006255687.2 | XP_006255749.1 | 3 - 4 | 420 | 98 | ||
XM_006255689.2 | XP_006255751.1 | 4 - 5 | 724 | 98 | ||
XM_006255690.2 | XP_006255752.1 | 4 - 5 | 510 | 98 | ||
GenBank mRNA | BC085839.1 | - | 3 - 4 | 388 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
