Assay Details
Gene Symbol: Cenpn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008366.1 | NP_001008367.1 | 7 - 8 | 744 | 79 | |
XM_006255686.2 | XP_006255748.1 | 7 - 8 | 858 | 79 | ||
XM_006255687.2 | XP_006255749.1 | 7 - 8 | 776 | 79 | ||
XM_006255689.2 | XP_006255751.1 | 8 - 9 | 1080 | 79 | ||
XM_006255690.2 | XP_006255752.1 | 8 - 9 | 866 | 79 | ||
XM_006255691.2 | XP_006255753.1 | 6 - 7 | 800 | 79 | ||
XM_006255692.2 | XP_006255754.1 | 6 - 7 | 680 | 79 | ||
GenBank mRNA | AY325257.1 | - | 866 | 79 | ||
AY325261.1 | - | 866 | 79 | |||
BC085839.1 | - | 7 - 8 | 744 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
