Assay Details
Gene Symbol: Wwox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106188.1 | NP_001099658.1 | 3 - 4 | 332 | 106 | |
XM_006255666.2 | XP_006255728.1 | 3 - 4 | 432 | 106 | ||
XM_006255670.2 | XP_006255732.1 | 3 - 4 | 428 | 106 | ||
XM_006255671.2 | XP_006255733.1 | 3 - 4 | 428 | 106 | ||
XM_006255673.2 | XP_006255735.1 | 3 - 4 | 428 | 106 | ||
XM_008772491.1 | XP_008770713.1 | 3 - 4 | 433 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
