Assay Details
Gene Symbol: Slc7a6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107424.1 | NP_001100894.1 | 1 - 2 | 553 | 123 | |
XM_006255498.2 | XP_006255560.1 | 2 - 3 | 618 | 123 | ||
XM_006255499.2 | XP_006255561.1 | 2 - 3 | 648 | 123 | ||
XM_006255500.2 | XP_006255562.1 | 2 - 3 | 766 | 123 | ||
XM_008772500.1 | XP_008770722.1 | 2 - 3 | 652 | 123 | ||
XM_008772501.1 | XP_008770723.1 | 2 - 3 | 833 | 123 | ||
XM_008772502.1 | XP_008770724.1 | 2 - 3 | 882 | 123 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
