Assay Details
Gene Symbol: Pla2g15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004277.2 | NP_001004277.1 | 3 - 4 | 433 | 67 | |
GenBank mRNA | AY490816.1 | - | 3 - 4 | 405 | 67 | |
BC098894.1 | - | 3 - 4 | 433 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
