Assay Details
Gene Symbol: Exoc3l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106178.1 | NP_001099648.1 | 10 - 11 | 1886 | 90 | |
XM_006255456.2 | XP_006255518.1 | 11 - 12 | 2303 | 90 | ||
XM_006255457.2 | XP_006255519.1 | 11 - 12 | 2639 | 90 | ||
XM_006255460.2 | XP_006255522.1 | 10 - 11 | 1817 | 90 | ||
XM_006255461.2 | XP_006255523.1 | 10 - 11 | 1766 | 90 | ||
XM_006255463.1 | XP_006255525.1 | 8 - 9 | 1511 | 90 | ||
XM_008772478.1 | XP_008770700.1 | 11 - 12 | 2522 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
