Assay Details
Gene Symbol: Exoc3l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106178.1 | NP_001099648.1 | 11 - 12 | 2000 | 107 | |
XM_006255456.2 | XP_006255518.1 | 12 - 13 | 2417 | 107 | ||
XM_006255457.2 | XP_006255519.1 | 12 - 13 | 2753 | 107 | ||
XM_006255460.2 | XP_006255522.1 | 11 - 12 | 1931 | 107 | ||
XM_006255461.2 | XP_006255523.1 | 11 - 12 | 1880 | 107 | ||
XM_006255463.1 | XP_006255525.1 | 9 - 10 | 1625 | 107 | ||
XM_008772478.1 | XP_008770700.1 | 12 - 13 | 2636 | 107 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
