Assay Details
Gene Symbol: Exoc3l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106178.1 | NP_001099648.1 | 12 - 13 | 2158 | 106 | |
XM_006255456.2 | XP_006255518.1 | 13 - 14 | 2575 | 106 | ||
XM_006255457.2 | XP_006255519.1 | 13 - 14 | 2911 | 106 | ||
XM_006255460.2 | XP_006255522.1 | 12 - 13 | 2089 | 106 | ||
XM_006255461.2 | XP_006255523.1 | 12 - 13 | 2038 | 106 | ||
XM_006255463.1 | XP_006255525.1 | 10 - 11 | 1783 | 106 | ||
XM_008772478.1 | XP_008770700.1 | 13 - 14 | 2794 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
