Assay Details
Gene Symbol: Bco1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053648.3 | NP_446100.2 | 10 - 11 | 1523 | 144 | |
GenBank mRNA | FQ219222.1 | - | 10 - 11 | 1574 | 144 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P222122 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898897 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83663 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 639626 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 880498 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 412732 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 274896 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 411426 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 656881 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 815246 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 675605 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 645073 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 638663 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 944723 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 403082 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 894108 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 849401 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 549192 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 273603 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404285 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 162155 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 448843 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 931616 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 801480 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 626966 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 103241 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 177200 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 125911 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 144231 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 125721 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P222122 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P222122 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P222122 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P222122 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P474388 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 23190 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 849 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 24568 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10808224 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808227 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808231 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808232 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808226 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808229 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808228 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808233 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808230 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808234 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808225 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10808223 | - | - |
Rat Genome 230 2.0 Array | 1369430_PM_at | - | - |
Rat Genome 230 2.0 Array | 1369430_at | - | - |
Affymetrix HT Rat Focus Array | 1369430_at | - | - |
Affymetrix HT Rat Focus Array | 1369430_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1900005655.rn.1 | TC1900000679.rn.1 | RTA TC |
Rat Expression Set 230 | 1369430_at | - | - |
Rat Expression Set 230 | 1369430_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10808223 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22376636 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1360882 | - | - |
RatRef-12 Expression BeadChip | ILMN_63594 | - | - |
