Assay Details
Gene Symbol: Psmb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025637.1 | NP_001020808.1 | 4 - 5 | 495 | 99 | |
GenBank mRNA | BC098835.1 | - | 4 - 5 | 495 | 99 | |
FQ226463.1 | - | 4 - 5 | 488 | 99 | ||
FQ228226.1 | - | 4 - 5 | 476 | 98 | ||
FQ229295.1 | - | 4 - 5 | 493 | 99 | ||
FQ229779.1 | - | 548 | 99 | |||
FQ233484.1 | - | 4 - 5 | 505 | 99 | ||
FQ235075.1 | - | 4 - 5 | 478 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
