Assay Details
Gene Symbol: Psmb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025637.1 | NP_001020808.1 | 7 - 8 | 828 | 80 | |
GenBank mRNA | BC098835.1 | - | 7 - 8 | 828 | 80 | |
FQ217478.1 | - | 3 - 4 | 371 | 80 | ||
FQ226463.1 | - | 7 - 8 | 821 | 80 | ||
FQ228226.1 | - | 7 - 8 | 808 | 80 | ||
FQ229295.1 | - | 7 - 8 | 826 | 80 | ||
FQ233484.1 | - | 7 - 8 | 838 | 80 | ||
FQ234080.1 | - | 7 - 8 | 830 | 80 | ||
FQ235075.1 | - | 7 - 8 | 811 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
