Assay Details
Gene Symbol: Nqo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017000.3 | NP_058696.2 | 1 - 2 | 94 | 81 | |
GenBank mRNA | BC083542.1 | - | 1 - 2 | 94 | 81 | |
J02608.1 | - | 1 - 2 | 79 | 81 | ||
J02679.1 | - | 1 - 2 | 79 | 81 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC083542.1, J02608.1, J02679.1, NM_017000.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
