Assay Details
Gene Symbol: Nqo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017000.3 | NP_058696.2 | 3 - 4 | 394 | 109 | |
GenBank mRNA | BC083542.1 | - | 3 - 4 | 394 | 109 | |
J02608.1 | - | 3 - 4 | 379 | 109 | ||
J02640.1 | - | 2 - 3 | 353 | 109 | ||
J02679.1 | - | 3 - 4 | 379 | 109 | ||
X17464.1 | - | 1 - 2 | 68 | 109 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
