Assay Details
Gene Symbol: Rnaseh2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013234.1 | NP_001013252.1 | 1 - 2 | 199 | 69 | |
GenBank mRNA | BC086539.1 | - | 1 - 2 | 199 | 69 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC086539.1, NM_001013234.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
