Assay Details
Gene Symbol: Cdh15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207613.1 | NP_997496.1 | 5 - 6 | 682 | 77 | |
GenBank mRNA | AB176538.1 | - | 5 - 6 | 682 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
