Assay Details
Gene Symbol: Dhodh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008553.1 | NP_001008553.1 | 5 - 6 | 762 | 103 | |
XM_006255601.2 | XP_006255663.1 | 5 - 6 | 935 | 103 | ||
XM_008772529.1 | XP_008770751.1 | 5 - 6 | 935 | 103 | ||
GenBank mRNA | X80778.1 | - | 5 - 6 | 762 | 103 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
