Assay Details
Gene Symbol: Ednra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012550.2 | NP_036682.1 | 1 - 2 | 236 | 111 | |
XM_006255420.2 | XP_006255482.1 | 1 - 2 | 402 | 111 | ||
XM_008772471.1 | XP_008770693.1 | 1 - 2 | 109 | 111 | ||
GenBank mRNA | CB798671.1 | - | 1 - 2 | 236 | 111 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_012550.2, XM_006255420.2, XM_008772471.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
