Assay Details
Gene Symbol: Abce1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108446.1 | NP_001101916.1 | 11 - 12 | 1268 | 94 | |
XM_006255407.2 | XP_006255469.1 | 11 - 12 | 1285 | 94 | ||
GenBank mRNA | AY724536.1 | - | 11 - 12 | 1217 | 94 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
