Assay Details
Gene Symbol: Abce1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108446.1 | NP_001101916.1 | 14 - 15 | 1503 | 76 | |
XM_006255407.2 | XP_006255469.1 | 14 - 15 | 1520 | 76 | ||
GenBank mRNA | AY724536.1 | - | 14 - 15 | 1452 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
