Assay Details
Gene Symbol: Cbfa2t3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108453.1 | NP_001101923.1 | 11 - 12 | 1679 | 63 | |
XM_006255770.2 | XP_006255832.1 | 10 - 11 | 2573 | 63 | ||
XM_006255772.2 | XP_006255834.1 | 11 - 12 | 1834 | 63 | ||
XM_008772629.1 | XP_008770851.1 | 10 - 11 | 3544 | 63 | ||
XM_008772630.1 | XP_008770852.1 | 9 - 10 | 3467 | 63 | ||
XM_008772631.1 | XP_008770853.1 | 10 - 11 | 2029 | 63 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription by RNA polymerase II
granulocyte differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription by RNA polymerase II
chromatin binding
DNA-binding transcription factor activity
transcription corepressor activity
metal ion binding
negative regulation of transcription by RNA polymerase II
granulocyte differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription by RNA polymerase II
chromatin binding
DNA-binding transcription factor activity
transcription corepressor activity
metal ion binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P118244 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 362729 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 674914 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 75211 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729432 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 573792 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147723 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 318275 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 369052 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323444 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 826615 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 378699 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 836137 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 818687 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 783047 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 635485 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 72218 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873053 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 457836 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 295108 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 738777 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 838035 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173874 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325401 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 198512 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 220507 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 670585 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 610399 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70856 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 726474 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 264111 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 678166 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 129479 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 812655 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 565711 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 3483 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 50525 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 279192 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 460093 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 607541 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 760735 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427199 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 426404 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 299464 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 406236 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 797902 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522515 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 862437 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 715897 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 885898 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 200339 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 547153 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 802475 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681500 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93641 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 126357 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 178644 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 52562 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 110881 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 67504 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 41529 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 104059 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 115220 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 925 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 167421 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 57135 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 124689 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 149480 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P118244 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P118244 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P118244 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P21717 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 15817 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 72192 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10811699 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10811691 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10811696 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10811698 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10811694 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10811689 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10811693 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10811692 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10811690 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10811701 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10811697 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10811700 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10811695 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10811688 | - | - |
Rat Genome 230 2.0 Array | 1377199_PM_at | - | - |
Rat Genome 230 2.0 Array | 1398231_PM_at | - | - |
Rat Genome 230 2.0 Array | 1378921_PM_at | - | - |
Rat Genome 230 2.0 Array | 1379674_at | - | - |
Rat Genome 230 2.0 Array | 1377199_at | - | - |
Rat Genome 230 2.0 Array | 1379674_PM_at | - | - |
Rat Genome 230 2.0 Array | 1398231_at | - | - |
Rat Genome 230 2.0 Array | 1378921_at | - | - |
Rat Genome 230 2.0 Array | 1392645_PM_at | - | - |
Rat Genome 230 2.0 Array | 1392645_at | - | - |
Affymetrix HT Rat Focus Array | 1379674_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1379674_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1900013212.rn.1 | TC1900001641.rn.1 | RTA TC |
Rat Expression Set 230 | 1379674_at | - | - |
Rat Expression Set 230 | 1379674_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10811688 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20740607 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1369167 | - | - |
