Assay Details
Gene Symbol: Cbfa2t3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108453.1 | NP_001101923.1 | 9 - 10 | 1423 | 86 | |
XM_006255770.2 | XP_006255832.1 | 8 - 9 | 2317 | 86 | ||
XM_006255772.2 | XP_006255834.1 | 9 - 10 | 1578 | 86 | ||
XM_008772629.1 | XP_008770851.1 | 8 - 9 | 3288 | 86 | ||
XM_008772630.1 | XP_008770852.1 | 7 - 8 | 3211 | 86 | ||
XM_008772631.1 | XP_008770853.1 | 8 - 9 | 1773 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
