Assay Details
Gene Symbol: Plaa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053866.3 | NP_446318.3 | 11 - 12 | 1666 | 69 | |
GenBank mRNA | CF110238.1 | - | 1 - 2 | 37 | 69 | |
CV110320.1 | - | 4 - 5 | 526 | 69 | ||
FQ211509.1 | - | 1727 | 69 | |||
FQ223782.1 | - | 11 - 12 | 1741 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
