Assay Details
Gene Symbol: Pgm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017033.2 | NP_058729.2 | 1 - 2 | 313 | 87 | |
GenBank mRNA | BC099807.1 | - | 1 - 2 | 313 | 87 | |
BC128703.1 | - | 1 - 2 | 318 | 87 | ||
L11694.1 | - | 1 - 2 | 296 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
