Assay Details
Gene Symbol: Alg6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033709.1 | NP_001028881.1 | 12 - 13 | 1327 | 147 | |
GenBank mRNA | BC101850.1 | - | 12 - 13 | 1327 | 147 | |
FQ210318.1 | - | 12 - 13 | 1342 | 147 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
