Assay Details
Gene Symbol: Angptl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025065.1 | NP_001020236.1 | 4 - 5 | 874 | 81 | |
XM_006238440.2 | XP_006238502.1 | 4 - 5 | 1061 | 81 | ||
GenBank mRNA | BC088192.1 | - | 4 - 5 | 874 | 81 | |
FQ210243.1 | - | 4 - 5 | 887 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
