Assay Details
Gene Symbol: Tek | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105737.1 | NP_001099207.1 | 13 - 14 | 2531 | 154 | |
XM_008763910.1 | XP_008762132.1 | 13 - 14 | 2351 | 154 | ||
GenBank mRNA | AF030423.1 | - | 1 - 2 | 154 | 154 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
