Assay Details
Gene Symbol: Tek | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105737.1 | NP_001099207.1 | 14 - 15 | 2686 | 64 | |
XM_008763910.1 | XP_008762132.1 | 14 - 15 | 2506 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
