Assay Details
Gene Symbol: Gprc6a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271106.1 | NP_001258035.1 | 2 - 3 | 522 | 88 | |
GenBank mRNA | AJ535460.1 | - | 2 - 3 | 498 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
