Assay Details
Gene Symbol: Lims1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006256385.2 | XP_006256447.1 | 1 - 2 | 2072 | 84 | |
XM_006256386.2 | XP_006256448.1 | 1 - 2 | 2072 | 84 | ||
XM_006256387.2 | XP_006256449.1 | 1 - 2 | 2069 | 84 | ||
GenBank mRNA | FJ640985.1 | - | 1 - 2 | 249 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
