Assay Details
Gene Symbol: Pkib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271158.1 | NP_001258087.1 | 3 - 4 | 203 | 107 | |
NM_012627.3 | NP_036759.2 | 3 - 4 | 263 | 107 | ||
GenBank mRNA | BC078880.1 | - | 3 - 4 | 203 | 107 | |
FQ231722.1 | - | 3 - 4 | 259 | 107 | ||
M64092.1 | - | 3 - 4 | 220 | 107 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC078880.1, M64092.1, NM_001271158.1, NM_012627.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
