Assay Details
Gene Symbol: Aifm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006256452.2 | XP_006256514.1 | 9 - 10 | 1284 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
