Assay Details
Gene Symbol: Acyp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169145.1 | NP_001162616.1 | 1 - 2 | 270 | 106 | |
XM_008770465.1 | XP_008768687.1 | 1 - 2 | 368 | 106 | ||
XM_008770466.1 | XP_008768688.1 | 1 - 2 | 351 | 106 | ||
GenBank mRNA | CB613181.1 | - | 1 - 2 | 280 | 106 | |
CO404339.1 | - | 1 - 2 | 101 | 106 | ||
FQ215015.1 | - | 1 - 2 | 102 | 106 | ||
FQ224926.1 | - | 1 - 2 | 138 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
