Assay Details
Gene Symbol: Acyp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169145.1 | NP_001162616.1 | 3 - 4 | 402 | 101 | |
GenBank mRNA | CB613181.1 | - | 3 - 4 | 412 | 101 | |
CO404339.1 | - | 3 - 4 | 233 | 101 | ||
FQ215015.1 | - | 3 - 4 | 234 | 101 | ||
FQ224926.1 | - | 3 - 4 | 270 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
