Assay Details
Gene Symbol: Cct4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182814.2 | NP_877966.1 | 6 - 7 | 779 | 78 | |
GenBank mRNA | AY223861.1 | - | 6 - 7 | 645 | 78 | |
BC079283.1 | - | 6 - 7 | 750 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
