Assay Details
Gene Symbol: Dusp22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108412.1 | NP_001101882.1 | 3 - 4 | 260 | 76 | |
XM_006253888.2 | XP_006253950.1 | 3 - 4 | 208 | 76 | ||
XM_006253889.2 | XP_006253951.1 | 3 - 4 | 481 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
