Assay Details
Gene Symbol: Dusp22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108412.1 | NP_001101882.1 | 7 - 8 | 628 | 125 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription by RNA polymerase II
negative regulation of T cell mediated immunity
protein dephosphorylation
transforming growth factor beta receptor signaling pathway
peptidyl-tyrosine dephosphorylation
regulation of cell population proliferation
positive regulation of JNK cascade
negative regulation of T cell receptor signaling pathway
negative regulation of T cell activation
protein tyrosine phosphatase activity
protein tyrosine/serine/threonine phosphatase activity
negative regulation of transcription by RNA polymerase II
negative regulation of T cell mediated immunity
protein dephosphorylation
transforming growth factor beta receptor signaling pathway
peptidyl-tyrosine dephosphorylation
regulation of cell population proliferation
positive regulation of JNK cascade
negative regulation of T cell receptor signaling pathway
negative regulation of T cell activation
protein tyrosine phosphatase activity
protein tyrosine/serine/threonine phosphatase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P974489 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 931975 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 461796 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 216099 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 533642 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 661648 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 100727 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 239004 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 402363 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 311824 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 237914 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427973 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 8488 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 105855 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 454952 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 831639 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 546513 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284446 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77778 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 534961 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 256472 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 748104 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 207970 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 186692 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 455800 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 333469 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 320684 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 445928 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 840697 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452532 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 406641 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 868947 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 761234 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 334634 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 440088 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 846274 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 834274 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 42657 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59653 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 127150 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33942 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 170328 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P974489 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P974489 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P974489 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P403108 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P974489 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 97408 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 63143 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10798187 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10798185 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10798188 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10798192 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10798193 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10798190 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10798189 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10798191 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10798184 | - | - |
Rat Genome 230 2.0 Array | 1375967_a_at | - | - |
Rat Genome 230 2.0 Array | 1375967_PM_a_at | - | - |
Affymetrix HT Rat Focus Array | 1375967_PM_a_at | - | - |
Affymetrix HT Rat Focus Array | 1375967_a_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1700011692.rn.1 | TC1700001700.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1700011696.rn.1 | TC1700001700.rn.1 | RTA TC |
Rat Expression Set 230 | 1375967_PM_a_at | - | - |
Rat Expression Set 230 | 1375967_a_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10798184 | - | - |
Rat Genome U34 Set | rc_AI171617_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21634747 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1369425 | - | - |
