Assay Details
Gene Symbol: Mboat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109120.1 | NP_001102590.1 | 2 - 3 | 430 | 80 | |
GenBank mRNA | BC168934.1 | - | 2 - 3 | 387 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
