Assay Details
Gene Symbol: Ptpdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106104.1 | NP_001099574.1 | 5 - 6 | 715 | 108 | |
XM_006253700.2 | XP_006253762.1 | 4 - 5 | 1538 | 108 | ||
XM_006253701.2 | XP_006253763.1 | 5 - 6 | 1235 | 108 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
