Assay Details
Gene Symbol: Ptpdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106104.1 | NP_001099574.1 | 8 - 9 | 2292 | 80 | |
XM_006253700.2 | XP_006253762.1 | 7 - 8 | 3115 | 80 | ||
XM_006253701.2 | XP_006253763.1 | 8 - 9 | 2812 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
