Assay Details
Gene Symbol: Ippk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008556.1 | NP_001008556.1 | 10 - 11 | 1060 | 105 | |
XM_008771534.1 | XP_008769756.1 | 9 - 10 | 1775 | 105 | ||
XM_008771535.1 | XP_008769757.1 | 10 - 11 | 1159 | 105 | ||
GenBank mRNA | AY823319.1 | - | 10 - 11 | 1060 | 105 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
