Assay Details
Gene Symbol: Gmpr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057188.1 | NP_476536.1 | 8 - 9 | 931 | 81 | |
GenBank mRNA | AF090867.1 | - | 8 - 9 | 931 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P409518 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 364229 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468071 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 207370 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 216144 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14613 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 736733 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 928676 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 556623 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669725 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 873937 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839491 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 80079 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 641468 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 970011 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 459975 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 512915 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732119 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 745214 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158524 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 358815 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 378489 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 773733 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 310945 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789975 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 414903 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 139129 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 345168 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 118667 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 26189 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 9741 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 23780 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 101280 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 23256 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 29313 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P409518 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P409518 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P409518 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P409518 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 76824 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 25626 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 68593 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10797787 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10797794 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10797791 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10797793 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10797789 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10797790 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10797792 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10797788 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10797795 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10797786 | - | - |
Rat Genome 230 2.0 Array | 1386914_PM_at | - | - |
Rat Genome 230 2.0 Array | 1386914_at | - | - |
Affymetrix HT Rat Focus Array | 1386914_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1386914_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1700010460.rn.1 | TC1700001499.rn.1 | RTA TC |
Rat Expression Set 230 | 1386914_PM_at | - | - |
Rat Expression Set 230 | 1386914_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10797786 | - | - |
Rat Genome U34 Set | AF090867_at | - | - |
Rat Genome U34 Set | AF090867_g_at | - | - |
Rat Genome U34 Set | AF090867_g_at | - | - |
Rat Genome U34 Set | AF090867_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21741167 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1371475 | - | - |
RatRef-12 Expression BeadChip | ILMN_61568 | - | - |
