Assay Details
Gene Symbol: Slc22a23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022624.1 | NP_072146.1 | 2 - 3 | 1502 | 112 | |
XM_006253845.2 | XP_006253907.1 | 2 - 3 | 782 | 112 | ||
GenBank mRNA | BC158843.1 | - | 2 - 3 | 401 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
