Assay Details
Gene Symbol: Eci3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009275.1 | NP_001009275.1 | 8 - 9 | 882 | 81 | |
GenBank mRNA | BC088178.1 | - | 8 - 9 | 882 | 81 | |
FQ209521.1 | - | 7 - 8 | 722 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
