Assay Details
Gene Symbol: Pak1ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037356.1 | NP_001032433.1 | 6 - 7 | 696 | 116 | |
GenBank mRNA | BC079236.1 | - | 6 - 7 | 676 | 116 | |
BC089822.1 | - | 7 - 8 | 757 | 116 | ||
BC107922.1 | - | 6 - 7 | 696 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
