Assay Details
Gene Symbol: Pak1ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037356.1 | NP_001032433.1 | 9 - 10 | 1026 | 63 | |
GenBank mRNA | BC079236.1 | - | 9 - 10 | 1006 | 63 | |
BC089822.1 | - | 10 - 11 | 1087 | 63 | ||
BC107922.1 | - | 9 - 10 | 1026 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
