Assay Details
Gene Symbol: Rabepk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024871.1 | NP_001020042.1 | 4 - 5 | 647 | 89 | |
XM_006233955.1 | XP_006234017.1 | 3 - 4 | 694 | 89 | ||
XM_006233957.2 | XP_006234019.1 | 3 - 4 | 302 | 89 | ||
XM_006233958.1 | XP_006234020.1 | 3 - 4 | 561 | 89 | ||
XM_006233959.2 | XP_006234021.1 | 3 - 4 | 372 | 89 | ||
XM_008761724.1 | XP_008759946.1 | 5 - 6 | 844 | 89 | ||
XM_008761725.1 | XP_008759947.1 | 4 - 5 | 402 | 89 | ||
XM_008761726.1 | XP_008759948.1 | 4 - 5 | 375 | 89 | ||
XM_008761727.1 | XP_008759949.1 | 3 - 4 | 721 | 89 | ||
XM_008761728.1 | XP_008759950.1 | 3 - 4 | 330 | 89 | ||
XM_008761729.1 | XP_008759951.1 | 3 - 4 | 588 | 89 | ||
XM_008761730.1 | XP_008759952.1 | 4 - 5 | 646 | 89 | ||
XM_008761733.1 | XP_008759955.1 | 2 - 3 | 612 | 89 | ||
XM_008761736.1 | XP_008759958.1 | 2 - 3 | 264 | 89 | ||
GenBank mRNA | BC097415.1 | - | 4 - 5 | 647 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
