Assay Details
Gene Symbol: Brd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108575.1 | NP_001102045.1 | 5 - 6 | 916 | 107 | |
XM_006233825.2 | XP_006233887.1 | 5 - 6 | 919 | 107 | ||
XM_006233826.2 | XP_006233888.1 | 5 - 6 | 945 | 107 | ||
XM_006233830.2 | XP_006233892.1 | 5 - 6 | 919 | 107 | ||
GenBank mRNA | BC168949.1 | - | 5 - 6 | 864 | 107 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
