Assay Details
Gene Symbol: Brd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108575.1 | NP_001102045.1 | 8 - 9 | 1606 | 93 | |
XM_006233830.2 | XP_006233892.1 | 8 - 9 | 1609 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
