Assay Details
Gene Symbol: Hspa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013083.2 | NP_037215.1 | 2 - 3 | 302 | 62 | |
GenBank mRNA | BC062017.1 | - | 2 - 3 | 262 | 62 | |
CB578015.1 | - | 2 - 3 | 302 | 62 | ||
M14050.1 | - | 1 - 2 | 178 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
