Assay Details
Gene Symbol: Ntng2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006233804.2 | XP_006233866.1 | 4 - 5 | 1287 | 133 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
